missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Cerium(IV) sulfate hydrate, 98%

Manufacturer:  Alfa Aesar™ A1014230

 View more versions of this product

Catalog No. AAA1014230



Description

Description

Applications
Cerium(IV) sulfate hydrate is used as a general application for chemical intermediate.

Solubility
Soluble in water.

Notes
Store in cool, dry conditions in well sealed containers. keep container tightly closed
Specifications

Specifications

Cerium (IV) sulfate hydrate
CeH2O9S2
UN3085
sulfuricacid, cerium salt, hydrate 9ci, acmc-20mqih, cerium iv sulfate monohydrate, cerium iv sulfate hydrate, reacton, cerium 4+ sulfate-water 1/2/1
O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Ce+4]
350.243
332.24 (Anhydrous)
195°C (decomposition)
95838-16-7
MFCD00149427
14,1990
KNANFSXYDCURQS-UHFFFAOYSA-J
cerium(4+);disulfate;hydrate
20223355
98%
250g

Chemical Identifiers

95838-16-7
350.243
KNANFSXYDCURQS-UHFFFAOYSA-J
20223355
O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Ce+4]
CeH2O9S2
MFCD00149427
sulfuricacid, cerium salt, hydrate 9ci, acmc-20mqih, cerium iv sulfate monohydrate, cerium iv sulfate hydrate, reacton, cerium 4+ sulfate-water 1/2/1
cerium(4+);disulfate;hydrate
Safety and Handling

Safety and Handling

GHS H Statement
H314-H272
Causes severe skin burns and eye damage.
May intensify fire
oxidizer.

GHS P Statement
P221-P210-P303+P361+P353-P305+P351+P338-P405-P501a
Take any precaution to avoid mixing with combustibles.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: OXIDIZING SOLID, CORROSIVE, N.O.S.

RTECSNumber : WS6960000

TSCA : TSCA

SDS Documents