Please login to your online account to display your discounted pricing

Alfa Aesar™ Cerium(IV) sulfate, anhydrous, 97%

$64.65 - $64.65

Chemical Identifiers

Chemical Name or Material Cerium(IV) sulfate
CAS 13590-82-4
Name Note anhydrous
Molecular Formula Ce(SO4)2
Formula Weight 332.24
MDL Number MFCD00148852
UN Number UN3085
Merck Index 14,1990
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL2025422

 
alfa aesar™
L2025422
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL2025414

 
alfa aesar™
L2025414
25g Each for $64.65

AAL2025436

 
alfa aesar™
L2025436
500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 332.24
Merck Index 14,1990
Quantity 100g
Health Hazard 3 GHS P Statement
P221-P210-P303+P361+P353-P305+P351+P338-P405-P501a
Take any precaution to avoid mixing with combustibles.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material Cerium(IV) sulfate
CAS 13590-82-4
Health Hazard 1 Danger
TSCA TSCA
Name Note anhydrous
Assay 97%
Molecular Formula Ce(SO4)2
MDL Number MFCD00148852
UN Number UN3085
DOT Information Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: OXIDIZING SOLID, CORROSIVE, N.O.S.
RTECS Number WS6960000
Health Hazard 2 GHS H Statement
H314-H272
Causes severe skin burns and eye damage.
May intensify fire; oxidizer.

Applications
Cerium(IV) sulfate is a redox indicator and is used in analytical chemistry for redox titrations. In ionized form, ceric ion acts as a strong oxidizer under acidic conditions. It is also used to prepare spherical-like nanoparticles CeO2, nanoparticles for NO reductions′ reactions.

Solubility
Soluble in water and dilute sulfuric acid.

Notes
Store in cool place. Hygroscopic and moisture sensitive. Avoid moisture. Incompatible with strong reducing agents.

Safety and Handling

Danger

GHS P Statement
P221-P210-P303+P361+P353-P305+P351+P338-P405-P501a
Take any precaution to avoid mixing with combustibles.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H272
Causes severe skin burns and eye damage.
May intensify fire; oxidizer.