Please login to your online account to display your discounted pricing

Cerium(IV) oxide, REacton™, 99.9% (REO), Alfa Aesar™

Chemical Identifiers

Chemical Name or Material Cerium(IV) oxide, REacton™
CAS 1306-38-3
Name Note (REO)
Molecular Formula CeO2
Formula Weight 172.12
MDL Number MFCD00010933
Merck Index 14,1989
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AA11328A1

 
alfa aesar
11328A1
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 172.12
Quantity 1kg
Chemical Name or Material Cerium(IV) oxide, REacton™
TSCA TSCA
Odor Odorless
Name Note (REO)
Physical Form 5μm Powder
Assay 99.9%
Molecular Formula CeO2
MDL Number MFCD00010933
CAS 1306-38-3
Merck Index 14,1989
Color Pale Yellow
Melting Point 2600°C
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302
Harmful if swallowed.
Health Hazard 3 GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Density 7.132
RTECS Number FK6310000
Safety and Handling

Warning

GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302
Harmful if swallowed.