Please login to your online account to display your discounted pricing

Alfa Aesar™ Cerium(IV) oxide, polishing compound, 57g (2oz)

Polishing compound for IR crystal windows

Chemical Identifiers

Chemical Name or Material Cerium(IV) oxide
CAS 1306-38-3
Name Note Polishing compound
Formula Weight 172.12
MDL Number MFCD00010933
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA39802LA

  SDS
alfa aesar™
39802LA
57g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 172.12
Quantity 57g
Health Hazard 3 GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material Cerium(IV) oxide
TSCA TSCA
Name Note Polishing compound
Physical Form Particles
MDL Number MFCD00010933
CAS 1306-38-3
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302
Harmful if swallowed.
Odor Odorless
Particle Size APS ≈3 Micron
Weight 57g (2oz)
Safety and Handling

Warning

GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302
Harmful if swallowed.