Please login to your online account to display your discounted pricing

Alfa Aesar™ Cerium(IV) oxide pellets, 99.9% (metals basis)

Sintered pellets suitable for vacuum deposition

Chemical Identifiers

Chemical Name or Material Cerium(IV) oxide pellets
CAS 1306-38-3
Name Note approx. 9mm (0.35 in.) dia. x 5mm (0.20 in.) thick
Formula Weight 172.12
MDL Number MFCD00010933
Merck Index 14,1989
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4065022

 
alfa aesar™
4065022
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 172.12
Health Hazard 3 GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material Cerium(IV) oxide pellets
TSCA TSCA
Odor Odorless
Name Note approx. 9mm (0.35 in.) dia. x 5mm (0.20 in.) thick
Assay 99.9%
Assay Percent Notes (metals basis)
MDL Number MFCD00010933
CAS 1306-38-3
Merck Index 14,1989
Color Pale yellow
Melting Point 2600°C
Quantity 100g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302
Harmful if swallowed.
RTECS Number FK6310000
Safety and Handling

Warning

GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302
Harmful if swallowed.