Please login to your online account to display your discounted pricing

Alfa Aesar™ Cerium(IV) oxide, doped with 18% zirconium, 13% in H2O, nanoparticle dispersion, high pH, <5.0nm APS Number Weighted

Chemical Identifiers

Chemical Name or Material Cerium(IV) oxide
CAS 1306-38-3
Name Note Doped with 18% zirconium, 13% in H2O, nanoparticle dispersion
MDL Number MFCD00010933
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4725922

  SDS
alfa aesar™
4725922
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA4725936

  SDS
alfa aesar™
4725936
500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

pH High
Chemical Name or Material Cerium(IV) oxide
TSCA TSCA
Odor Odorless
Name Note Doped with 18% zirconium, 13% in H2O, nanoparticle dispersion
Particle Size <5.0nm APS Number Weighted
Physical Form Dispersion
MDL Number MFCD00010933
CAS 1306-38-3
Color Red-brown
Quantity 100g