missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cerium(IV) oxide, Alfa Aesar™

Manufacturer:  Alfa Aesar 4381809

 View more versions of this product

Catalog No. AA4381809



Specifications

Specifications

Cerium(IV) oxide
CeO2
ceric oxide, cerium dioxide, cerium iv oxide, ceria, cerium oxide, ceric dioxide, cerium 4+ oxide, needlal, nidoral, opaline
O=[Ce]=O
172.114
CHEBI:79089
7.132g/mL
1306-38-3
MFCD00010933
CETPSERCERDGAM-UHFFFAOYSA-N
dioxocerium
73963
172.12
10g

Chemical Identifiers

1306-38-3
172.114
CETPSERCERDGAM-UHFFFAOYSA-N
73963
dioxocerium
CeO2
MFCD00010933
ceric oxide, cerium dioxide, cerium iv oxide, ceria, cerium oxide, ceric dioxide, cerium 4+ oxide, needlal, nidoral, opaline
CHEBI:79089
O=[Ce]=O
Safety and Handling

Safety and Handling

EINECSNumber : 215-150-4

Documents