missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cerium(IV) oxide, Alfa Aesar™

$50.43 - $203.55

Chemical Identifiers

CAS 1306-38-3
Molecular Formula CeO2
Molecular Weight (g/mol) 172.114
MDL Number MFCD00010933
InChI Key CETPSERCERDGAM-UHFFFAOYSA-N
Synonym ceria
PubChem CID 73963
ChEBI CHEBI:79089
IUPAC Name dioxocerium
SMILES O=[Ce]=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA4381809 View Documents Alfa Aesar
4381809
10g Each for $50.43
 
AA4381818 View Documents Alfa Aesar
4381818
50g Each for $203.55
 
Specifications

Chemical Identifiers

1306-38-3
172.114
CETPSERCERDGAM-UHFFFAOYSA-N
73963
dioxocerium
CeO2
MFCD00010933
ceria
CHEBI:79089
O=[Ce]=O

Specifications

1306-38-3
MFCD00010933
CETPSERCERDGAM-UHFFFAOYSA-N
dioxocerium
73963
172.12
10g
CeO2
ceria
O=[Ce]=O
172.114
CHEBI:79089
7.132g/mL
Cerium(IV) oxide
Safety and Handling

Safety and Handling

EINECSNumber : 215-150-4