Please login to your online account to display your discounted pricing

Cerium(IV) oxide, 99.9%, (trace metal basis), ACROS Organics™


Quantity 500g
Packaging Plastic bottle

Manufacturer: fisher scientific acros  199125000

 View more versions of this product

Catalog No. AC199125000

  • / Each
 

Description & Specifications

Chemical Identifiers

Purity Grade Trace Metal Basis
Physical Form Powder
Formula Weight 172.12
Color White to yellow
Quantity 500g
EINECS Number 215-150-4
TSCA TSCA
Density 7.1300g/mL
RTECS Number FK6310000
Chemical Name or Material Cerium(IV) oxide
Name Note 99.9%
Packaging Plastic bottle
Assay Percent Range 99.9%
Molecular Formula CeO2
Linear Formula CeO2
MDL Number MFCD00010933
CAS 1306-38-3
Total Impurity 0.1% max.
Synonym Cerium dioxide

Specifications

Chemical Name or Material Cerium(IV) oxide
Name Note 99.9%
CAS 1306-38-3
Purity Grade Trace Metal Basis
Assay Percent Range 99.9%
Molecular Formula CeO2
Linear Formula CeO2
Formula Weight 172.12
MDL Number MFCD00010933
Synonym Cerium dioxide