missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Cerium(IV) oxide, 99.9%, (trace metal basis), Thermo Scientific™
Supplier: Thermo Scientific Chemicals 199120025
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
For ultra trace analysisSpecifications
Cerium(IV) oxide | |
1306-38-3 | |
99.9% | |
CeO2 | |
06,99 | |
ceric oxide, cerium dioxide, cerium iv oxide, ceria, cerium oxide, ceric dioxide, cerium 4+ oxide, needlal, nidoral, opaline | |
CETPSERCERDGAM-UHFFFAOYSA-N | |
dioxocerium | |
73963 | |
172.12 | |
Trace Metal Basis | |
7.13 |
99.9% | |
2.5kg | |
CeO2 | |
MFCD00010933 | |
15,1988 | |
Solubility in water: insoluble. Other solubilities: soluble in acids | |
O=[Ce]=O | |
172.12 | |
CHEBI:79089 | |
99.9% | |
Plastic bottle | |
7.1300g/mL |
Safety and Handling
EINECSNumber : 215-150-4
RTECSNumber : FK6310000
TSCA : TSCA
RUO â Research Use Only