missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Cerium(IV) oxide, 99.5% (REO)

Manufacturer:  Alfa Aesar™ 1292522

 View more versions of this product

Catalog No. AA1292522



Specifications

Specifications

Cerium(IV) oxide
(REO)
MFCD00010933
ceric oxide, cerium dioxide, cerium iv oxide, ceria, cerium oxide, ceric dioxide, cerium 4+ oxide, needlal, nidoral, opaline
O=[Ce]=O
172.114
CHEBI:79089
Powder
7.132g/mL
Yellow
100g
1306-38-3
CeO2
14,1989
CETPSERCERDGAM-UHFFFAOYSA-N
dioxocerium
73963
172.12
99.5%
Odorless
2600°C

Chemical Identifiers

1306-38-3
172.114
CETPSERCERDGAM-UHFFFAOYSA-N
73963
dioxocerium
CeO2
MFCD00010933
ceric oxide, cerium dioxide, cerium iv oxide, ceria, cerium oxide, ceric dioxide, cerium 4+ oxide, needlal, nidoral, opaline
CHEBI:79089
O=[Ce]=O
Safety and Handling

Safety and Handling

GHS H Statement
H302
Harmful if swallowed.

GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : FK6310000

TSCA : TSCA

Documents