missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Cerium(IV) oxide, 20% in H2O, nanoparticle dispersion, low pH, <5.0nm APS Number Weighted

Manufacturer:  Alfa Aesar™ 4723222

 View more versions of this product

Catalog No. AA4723222



Specifications

Specifications

Cerium(IV) oxide
1306-38-3
MFCD00010933
CETPSERCERDGAM-UHFFFAOYSA-N
dioxocerium
73963
172.12
1.23g/mL
<5.0nm APS Number Weighted
4.0
20% in H2O, nanoparticle dispersion
CeO2
ceric oxide, cerium dioxide, cerium iv oxide, ceria, cerium oxide, ceric dioxide, cerium 4+ oxide, needlal, nidoral, opaline
O=[Ce]=O
172.114
CHEBI:79089
Liquid
Odorless
Red to Brown
100g

Chemical Identifiers

1306-38-3
172.114
CETPSERCERDGAM-UHFFFAOYSA-N
73963
dioxocerium
CeO2
MFCD00010933
ceric oxide, cerium dioxide, cerium iv oxide, ceria, cerium oxide, ceric dioxide, cerium 4+ oxide, needlal, nidoral, opaline
CHEBI:79089
O=[Ce]=O
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H335
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P304+P340-P362-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Take off contaminated clothing and wash before reuse.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : FK6310000

TSCA : TSCA

SDS Documents