missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cerium(IV) oxide, 20% in H{2}O, nanoparticle dispersion, high pH, <5.0nm APS Number Weighted, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 04724036

 View more versions of this product

Catalog No. AA4724036



Description

Description

The addition of CeO2 to CuO/TiO2 increases specific surface area and inhibits sintering and pore size growth at high temperatures. It is used as an additive in glass industry. Cerium dioxide is mainly used as such or in formulation as a high precision polishing agent for glass products, as catalyst or catalyst support. Polishing and decolorizing glass; opacifier in vitreous enamels and photochromic glasses; heat-resistant alloy coatings; as a cracking catalyst; as a catalyst for automobile emission control; in ceramic coatings; in phosphors; in cathodes; in capacitors; in semiconductors; in refractory oxides; gemstone polishing, as raw material for the production of glass and ceramics as well as additives in paints and varnishes .

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Cerium(IV) oxide
1306-38-3
8.0
500 g
MFCD00010933
Very low water solubility (<0.123 ?g/L at 20°C) and soluble in concentrated mineral acid.
O=[Ce]=O
172.114
CHEBI:79089
Odorless
20% in H2O, nanoparticle dispersion
Red to Brown
Liquid
CeO2
ceric oxide, cerium dioxide, cerium iv oxide, ceria, cerium oxide, ceric dioxide, cerium 4+ oxide, needlal, nidoral, opaline
CETPSERCERDGAM-UHFFFAOYSA-N
dioxocerium
73963
172.12
1.23 g/mL

Chemical Identifiers

1306-38-3
172.114
CETPSERCERDGAM-UHFFFAOYSA-N
73963
dioxocerium
CeO2
MFCD00010933
ceric oxide, cerium dioxide, cerium iv oxide, ceria, cerium oxide, ceric dioxide, cerium 4+ oxide, needlal, nidoral, opaline
CHEBI:79089
O=[Ce]=O
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H335
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P304+P340-P362-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Take off contaminated clothing and wash before reuse.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

EINECSNumber : 215-150-4

RTECSNumber : FK6310000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only