Please login to your online account to display your discounted pricing

Cerium(IV) oxide, 20% in H2O, colloidal dispersion in water with dispersant, NanoTek™, Alfa Aesar™

For Use As a Cerium Oxide Water Borne Dispersion Suitable For Use in Alkaline Side Polishing Slurries. Submicron Particle Size Makes For Good Removal Rate With Minimal Subsurface Damage

Chemical Identifiers

Chemical Name or Material Cerium(IV) oxide
Name Note CE-6080, 20% in H2O, colloidal dispersion with dispersant
CAS 1306-38-3
Physical Form Dry Powder
Molecular Formula CeO2
Formula Weight 172.12g/mol
MDL Number MFCD00010933
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4491022

 
alfa aesar
44910-22
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 100g
Chemical Name or Material Cerium(IV) oxide
Name Note CE-6080, 20% in H2O, colloidal dispersion with dispersant
CAS 1306-38-3
Physical Form Dry Powder
Molecular Formula CeO2
Formula Weight 172.12g/mol
MDL Number MFCD00010933
Health Hazard 1 H303 : May be harmful if swallowed.
Safety and Handling

H303 : May be harmful if swallowed.