Learn More
Cerium(IV) ammonium sulfate dihydrate, ACS, 94% min, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 04019430
Description
Ammonium cerium(IV) sulfate dihydrate is an oxidizing agent in organic synthesis and it is used as an analytical reagent, in thin-layer chromatography for the detection of anti-depressant drugs in human blood and urine samples. It is used to wet etch the Co catalyst and synthesis of carbon Nanotubes. It is used in medical and pharmaceutical industry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Cerium(IV) ammonium sulfate dihydrate | |
For analysis ACS | |
10378-47-9 | |
250 g | |
(NH4)4Ce(SO4)4·2H2O | |
Ammonium cerium sulfate | |
632.56 (596.53 Anhydrous) | |
Yellow crystalline |
130°C | |
Yellow | |
Crystalline | |
94% min. | |
MFCD00149417 | |
Soluble in water and insoluble in ethanol. | |
ACS |
Chemical Identifiers
10378-47-9 | |
(NH4)4Ce(SO4)4·2H2O | |
MFCD00149417 | |
Ammonium cerium sulfate |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335-H500
EINECSNumber : 231-567-4
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only