Please login to your online account to display your discounted pricing

Cerium(IV) ammonium nitrate, REacton™, 99.5% (REO), Alfa Aesar™

Chemical Identifiers

Chemical Name or Material Cerium(IV) ammonium nitrate, REacton™
CAS 16774-21-3
Name Note (REO)
Molecular Formula H8CeN8O18
Linear Formula (NH4)2Ce(NO3)6
Formula Weight 548.23
MDL Number MFCD00151121
UN Number UN1477
Merck Index 14,1992
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AA1133222

  SDS
alfa aesar
1133222
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 548.23
Quantity 100g
Health Hazard 2 GHS H Statement
H272-H315-H319-H335
May intensify fire; oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
DOT Information Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: NITRATES, INORGANIC, N.O.S.
UN Number UN1477
Merck Index 14,1992
Color Orange
Physical Form Crystalline
Health Hazard 1 Danger
Linear Formula (NH4)2Ce(NO3)6
TSCA TSCA
Odor Pungent
Name Note (REO)
CAS 16774-21-3
Assay 99.5%
Health Hazard 3 GHS P Statement
P221-P210-P220-P305+P351+P338-P405-P501a
Take any precaution to avoid mixing with combustibles.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Keep/Store away from clothing/combustible materials.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material Cerium(IV) ammonium nitrate, REacton™
MDL Number MFCD00151121
Molecular Formula H8CeN8O18
Safety and Handling

Danger

GHS P Statement
P221-P210-P220-P305+P351+P338-P405-P501a
Take any precaution to avoid mixing with combustibles.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Keep/Store away from clothing/combustible materials.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H272-H315-H319-H335
May intensify fire; oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.