missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cerium(IV) ammonium nitrate, ACS, 98.5% min, Thermo Scientific Chemicals

Analytical chemistry, as an oxidant for organic compounds, as a polymerization catalyst for olefins

Supplier:  Thermo Scientific Chemicals 03325422

 View more versions of this product

Catalog No. AA3325422



Description

Description

Analytical chemistry, as an oxidant for organic compounds, as a polymerization catalyst for olefins, and as a scavenger in the manufacture of azides. Cerium(IV) ammonium nitrate is widely utilized as an oxidant for many functional groups like alcohols, phenols and ethers as well as C-H bonds. It acts as a deprotection reagent for alcohol in organic synthesis. It is useful in the synthesis of heterocycles like quinoxaline derivatives which is used in dyes, organic semiconductors and DNA cleaving agents. It is an active component of chrome etchant, which is used in the production of photomasks and liquid crystal displays.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Cerium(IV) ammonium nitrate
Granules
98.5%
MFCD00151121
548.23
16774-21-3
100 g
(NH4)2Ce(NO3)6
Soluble in water and alcohol.
ACS

Chemical Identifiers

16774-21-3
(NH4)2Ce(NO3)6
MFCD00151121
Safety and Handling

Safety and Handling

P210-P220-P221-P234-P260-P264b-P270-P271-P272-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P370+P378q-P390-P501c

H272-H290-H302-H314-H317-H335

DOTInformation : Hazard Class: 5.1; Packaging Group: II

EINECSNumber : 240-827-6

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only