Please login to your online account to display your discounted pricing

Alfa Aesar™ Cerium(IV) 2-methoxyethoxide, 18-20% w/w in 2-methoxyethanol

Chemical Identifiers

Chemical Name or Material Cerium(IV) 2-methoxyethoxide
CAS 876107-33-4
Linear Formula Ce[OCH2CH2OCH3]4
Formula Weight 440.47
MDL Number MFCD00236530
UN Number UN1188
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4047106

  SDS
alfa aesar™
4047106
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA4047122

  SDS
alfa aesar™
4047122
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA4047114

  SDS
alfa aesar™
4047114
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 440.47
Health Hazard 2 GHS H Statement
H360-H226-H302-H312-H332
May damage fertility or the unborn child.
Flammable liquid and vapor.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Density 1.02g/mL
UN Number UN1188
Concentration 18 to 20% w/w in 2-methoxyethanol
Chemical Name or Material Cerium(IV) 2-methoxyethoxide
Linear Formula Ce[OCH2CH2OCH3]4
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ETHYLENE GLYCOL MONOMETHYL ETHER
Health Hazard 1 Danger
Quantity 5g
MDL Number MFCD00236530
Health Hazard 3 GHS P Statement
P201-P308+P313
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Color Yellow
CAS 876107-33-4
Physical Form Liquid
Sensitivity Moisture Sensitive
Safety and Handling

Danger

GHS P Statement
P201-P308+P313
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.

GHS H Statement
H360-H226-H302-H312-H332
May damage fertility or the unborn child.
Flammable liquid and vapor.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.