Please login to your online account to display your discounted pricing

Alfa Aesar™ Cerium iron oxide, 20% in H2O, nanoparticle dispersion, low pH, <5.0nm APS Number Weighted

Chemical Identifiers

Chemical Name or Material Cerium iron oxide
Name Note 20% in H2O, nanoparticle dispersion
MDL Number MFCD02687051
Synonym Cerium and iron mixed metal oxide in water at low pH
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4728322

  SDS
alfa aesar™
4728322
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA4728336

  SDS
alfa aesar™
4728336
500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

pH Low
Chemical Name or Material Cerium iron oxide
Health Hazard 3 GHS P Statement
P261-P280-P304+P340-P362-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Take off contaminated clothing and wash before reuse.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Color Red-brown
Density 1.23g/mL
Odor Petroleum-like
Physical Form Liquid
Health Hazard 2 GHS H Statement
H302-H315-H335
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
MDL Number MFCD02687051
Quantity 100g
Particle Size <5.0nm APS Number Weighted
Synonym Cerium and iron mixed metal oxide in water at low pH
TSCA TSCA
Avg. Mol. Wt. or Mol. Wt. Range CeO2:Fe2O3; =50:50 wt%
Health Hazard 1 Warning
Name Note 20% in H2O, nanoparticle dispersion
Safety and Handling

Warning

GHS P Statement
P261-P280-P304+P340-P362-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Take off contaminated clothing and wash before reuse.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H315-H335
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.