Please login to your online account to display your discounted pricing

Alfa Aesar™ Cerium iron oxide, 2.0% in octanoic acid and Kensol 50H, nanoparticle dispersion, <5.0nm APS Number Weighted

Chemical Identifiers

Chemical Name or Material Cerium iron oxide
Name Note Nanoparticle dispersion
MDL Number MFCD02687051
Synonym Cerium and iron mixed metal oxide in octanoic acid and Kensol 50H
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4725122

  SDS
alfa aesar™
4725122
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA4725136

  SDS
alfa aesar™
4725136
500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Cerium iron oxide
TSCA TSCA
Odor Petroleum-like
Name Note Nanoparticle dispersion
Particle Size <5.0nm APS Number Weighted
Physical Form Liquid
MDL Number MFCD02687051
Synonym Cerium and iron mixed metal oxide in octanoic acid and Kensol 50H
Color Pink
Quantity 100g
Avg. Mol. Wt. or Mol. Wt. Range CeO2:Fe2O3; =50:50 wt%
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H350-H318-H315
May cause cancer.
Causes serious eye damage.
Causes skin irritation.
Health Hazard 3 GHS P Statement
P280-P281-P305+P351+P338-P310-P405-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Concentration 2.0% in octanoic acid and Kensol 50H
Density 0.84g/mL
Safety and Handling

Danger

GHS P Statement
P280-P281-P305+P351+P338-P310-P405-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H350-H318-H315
May cause cancer.
Causes serious eye damage.
Causes skin irritation.