missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Cerium(III) sulfate octahydrate, REacton™, 99.999% (metals basis)

Manufacturer:  Alfa Aesar™ 4435730

 View more versions of this product

Catalog No. AA4435730



Specifications

Specifications

Cerium (III) sulfate octahydrate
(metals basis)
MFCD00149641
Soluble in water.
Crystalline
Reacton™
250g
10450-59-6
Ce2(SO4)3·8H2O
14,2002
712.55 (568.42 Anhydrous)
99.999%
2.886 g/mL

Chemical Identifiers

10450-59-6
Ce2(SO4)3·8H2O
MFCD00149641
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents