Learn More
Cerium(III) perchlorate hexahydrate, Reagent Grade, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 01165430
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Cerium(III) perchlorate hexahydrate | |
Crystalline | |
CeCl3O12 | |
UN1481 | |
LJBTWTBUIINKRU-UHFFFAOYSA-K | |
cerium(3+);triperchlorate | |
166978 | |
Reagent | |
Moisture sensitive |
14017-47-1 | |
250 g | |
MFCD00150205 | |
cerium 3+ perchlorate, cerium perchlorate, cerous perchlorate, cerium 3+ triperchlorate, perchloric acid, cerium 3+ salt, perchloric acid, cerium 3+ salt 3:1, acmc-1bs3z, cerium iii perchlorate solution, perchloric acid cerium iii salt, perchloric acid,cerium 3+ salt 3:1 | |
[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Ce+3] | |
438.454 | |
546.56 (438.47 Anhydrous) | |
Odorless |
Chemical Identifiers
14017-47-1 | |
438.454 | |
LJBTWTBUIINKRU-UHFFFAOYSA-K | |
166978 | |
[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Ce+3] |
CeCl3O12 | |
MFCD00150205 | |
cerium 3+ perchlorate, cerium perchlorate, cerous perchlorate, cerium 3+ triperchlorate, perchloric acid, cerium 3+ salt, perchloric acid, cerium 3+ salt 3:1, acmc-1bs3z, cerium iii perchlorate solution, perchloric acid cerium iii salt, perchloric acid,cerium 3+ salt 3:1 | |
cerium(3+);triperchlorate |
Safety and Handling
GHS H Statement
H272-H315-H319-H335
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P220-P221-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
H272-H315-H319-H335
DOTInformation : Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: PERCHLORATES, INORGANIC, N.O.S.
EINECSNumber : 237-836-2
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only