Learn More
Alfa Aesar™ Cerium(III) oxalate hydrate, REacton™, 99.989% (REO)
Manufacturer: Alfa Aesar™ 1322609
Specifications
Cerium(III) oxalate hydrate | |
(REO) | |
MFCD00150139 | |
14,2001 | |
OGGMXARXHWPYSA-UHFFFAOYSA-N | |
cerium;oxalic acid;hydrate | |
91998046 | |
Crystalline | |
Reacton™ | |
Hygroscopic |
15750-47-7 | |
C6H8Ce2O13 | |
UN3288 | |
cerium iii oxalate hydrate, cerium iii oxalatehydrate, cerium iii oxalate decahydrate | |
C(=O)(C(=O)O)O.C(=O)(C(=O)O)O.C(=O)(C(=O)O)O.O.[Ce].[Ce] | |
568.349 | |
544.29 (Anhydrous) | |
99.989% | |
Odorless | |
10g |
Chemical Identifiers
15750-47-7 | |
568.349 | |
OGGMXARXHWPYSA-UHFFFAOYSA-N | |
91998046 | |
C(=O)(C(=O)O)O.C(=O)(C(=O)O)O.C(=O)(C(=O)O)O.O.[Ce].[Ce] |
C6H8Ce2O13 | |
MFCD00150139 | |
cerium iii oxalate hydrate, cerium iii oxalatehydrate, cerium iii oxalate decahydrate | |
cerium;oxalic acid;hydrate |
Safety and Handling
GHS H Statement
H302-H312
Harmful if swallowed.
Harmful in contact with skin.
GHS P Statement
P280-P301+P312-P312-P363-P322-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wash contaminated clothing before reuse.
Specific measures (see label).
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.
TSCA : TSCA