Please login to your online account to display your discounted pricing

Alfa Aesar™ Cerium(III) oxalate hydrate, REacton™, 99.989% (REO)

Chemical Identifiers

Chemical Name or Material Cerium(III) oxalate hydrate
CAS 15750-47-7
Purity Grade Reacton™
Formula Weight 544.29 (Anhydrous)
MDL Number MFCD00150139
UN Number UN3288
Merck Index 14,2001
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1322609

  SDS
alfa aesar™
1322609
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1322618

  SDS
alfa aesar™
1322618
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1322630

  SDS
alfa aesar™
1322630
250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 544.29 (Anhydrous)
Chemical Name or Material Cerium(III) oxalate hydrate
TSCA TSCA
Odor Odorless
Purity Grade Reacton™
Physical Form Crystalline
Assay 99.989%
Sensitivity Hygroscopic
Assay Percent Notes (REO)
MDL Number MFCD00150139
CAS 15750-47-7
UN Number UN3288
Merck Index 14,2001
Quantity 10g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302-H312
Harmful if swallowed.
Harmful in contact with skin.
Health Hazard 3 GHS P Statement
P280-P301+P312-P312-P363-P322-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wash contaminated clothing before reuse.
Specific measures (see label).
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.
Safety and Handling

Warning

GHS P Statement
P280-P301+P312-P312-P363-P322-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wash contaminated clothing before reuse.
Specific measures (see label).
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H312
Harmful if swallowed.
Harmful in contact with skin.