missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Cerium(III) oxalate hydrate, REacton™, 99.989% (REO)

Manufacturer:  Alfa Aesar™ 1322630

 View more versions of this product

Catalog No. AA1322630



Specifications

Specifications

Cerium(III) oxalate hydrate
(REO)
MFCD00150139
14,2001
OGGMXARXHWPYSA-UHFFFAOYSA-N
cerium;oxalic acid;hydrate
91998046
Crystalline
Reacton™
Hygroscopic
15750-47-7
C6H8Ce2O13
UN3288
cerium iii oxalate hydrate, cerium iii oxalatehydrate, cerium iii oxalate decahydrate
C(=O)(C(=O)O)O.C(=O)(C(=O)O)O.C(=O)(C(=O)O)O.O.[Ce].[Ce]
568.349
544.29 (Anhydrous)
99.989%
Odorless
250g

Chemical Identifiers

15750-47-7
568.349
OGGMXARXHWPYSA-UHFFFAOYSA-N
91998046
C(=O)(C(=O)O)O.C(=O)(C(=O)O)O.C(=O)(C(=O)O)O.O.[Ce].[Ce]
C6H8Ce2O13
MFCD00150139
cerium iii oxalate hydrate, cerium iii oxalatehydrate, cerium iii oxalate decahydrate
cerium;oxalic acid;hydrate
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312
Harmful if swallowed.
Harmful in contact with skin.

GHS P Statement
P280-P301+P312-P312-P363-P322-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wash contaminated clothing before reuse.
Specific measures (see label).
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.

TSCA : TSCA

Documents