Learn More
Cerium(III) oxalate hydrate, REacton™, 99.9% (REO), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 02112222
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Cerium(III) oxalate hydrate | |
15750-47-7 | |
100 g | |
C6Ce2O12 | |
UN3288 | |
cerium iii oxalate hydrate, cerium iii oxalatehydrate, cerium iii oxalate decahydrate | |
[Ce+3].[Ce+3].[O-]C(=O)C([O-])=O.[O-]C(=O)C([O-])=O.[O-]C(=O)C([O-])=O | |
544.29 | |
544.29 (Anhydrous) | |
Reacton™ | |
Hygroscopic |
(decomposition) | |
Powder | |
(REO) | |
MFCD00013082 | |
14,2001 | |
ZMZNLKYXLARXFY-UHFFFAOYSA-H | |
cerium;oxalic acid;hydrate | |
91998046 | |
99.9% | |
Odorless |
Chemical Identifiers
15750-47-7 | |
544.29 | |
ZMZNLKYXLARXFY-UHFFFAOYSA-H | |
91998046 |
C6Ce2O12 | |
MFCD00013082 | |
cerium iii oxalate hydrate, cerium iii oxalatehydrate, cerium iii oxalate decahydrate | |
[Ce+3].[Ce+3].[O-]C(=O)C([O-])=O.[O-]C(=O)C([O-])=O.[O-]C(=O)C([O-])=O |
Safety and Handling
GHS H Statement
H302-H312
Harmful if swallowed.
Harmful in contact with skin.
P264b-P270-P280-P301+P312-P302+P352-P312-P330-P363-P501c
H302+H312
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only