Learn More
Cerium(III) nitrate hydrate, REacton™, 99.998% (metals basis), Thermo Scientific Chemicals
$262.58 - $760.08
Chemical Identifiers
Molecular Formula | Ce(NO3)3·xH2O (x ≈ 6-7) |
---|---|
MDL Number | MFCD00010926 |
Description
Cerium(III) nitrate hydrate is used as an analytical reagent. It acts as a catalyst in petrochemical industry and in the pharmaceutical industry. Further, it is a chemical that helps burn tissue recover in vitro.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
Ce(NO3)3·xH2O (x ≈ 6-7) | |
MFCD00010926 |
Specifications
White | |
25 g | |
Ce(NO3)3·xH2O (x ≈ 6-7) | |
Soluble in water,alcohol and acetone | |
Acidic | |
White crystalline | |
Cerium(III) nitrate hydrate, 150°C - 3H2O |
Crystalline | |
99.998% | |
MFCD00010926 | |
326.13 (Anhydrous) | |
Hygroscopic | |
1.67g/cm3 at 20°C |
Safety and Handling
P201-P202-P220-P221-P264b-P271-P280-P302+P352-P305+P351+P338-P308+P313-P310-P332+P313-P362-P370+P380+P375-P372-P501c
H272-H315-H318-H350
DOTInformation : DOT Class: 5.1, Packaging Group: III
EINECSNumber : 233-297-2
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only