Please login to your online account to display your discounted pricing

Cerium(III) nitrate hexahydrate, REacton™, 99.5% (REO), Alfa Aesar™

Separation of cerium from other rare earths, catalyst for hydrolysis of phosporic acid esters

Chemical Identifiers

Chemical Name or Material Cerium(III) nitrate hexahydrate, REacton™
CAS 10294-41-4
Name Note (REO)
Molecular Formula CeN3O9·6H2O
Linear Formula Ce(NO3)3·6H2O
Formula Weight 434.23 (326.13 Anhydrous)
MDL Number MFCD00149631
UN Number UN1477
Merck Index 14,2000
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AA1132936

 
alfa aesar
1132936
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 434.23 (326.13 Anhydrous)
Quantity 500g
Chemical Name or Material Cerium(III) nitrate hexahydrate, REacton™
TSCA TSCA
Odor Odorless
Name Note (REO)
Physical Form Crystalline Aggregates
Assay 99.5%
Sensitivity Hygroscopic
Molecular Formula CeN3O9·6H2O
Linear Formula Ce(NO3)3·6H2O
MDL Number MFCD00149631
CAS 10294-41-4
UN Number UN1477
Merck Index 14,2000
pH 3
Solubility Information Soluble in water, alcohol, acetone
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H272-H318-H335-H315
May intensify fire; oxidizer.
Causes serious eye damage.
May cause respiratory irritation.
Causes skin irritation.
Health Hazard 3 GHS P Statement
P221-P210-P220-P305+P351+P338-P405-P501a
Take any precaution to avoid mixing with combustibles.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Keep/Store away from clothing/combustible materials.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: NITRATES, INORGANIC, N.O.S.
RTECS Number FK6300000
Safety and Handling

Danger

GHS P Statement
P221-P210-P220-P305+P351+P338-P405-P501a
Take any precaution to avoid mixing with combustibles.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Keep/Store away from clothing/combustible materials.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H272-H318-H335-H315
May intensify fire; oxidizer.
Causes serious eye damage.
May cause respiratory irritation.
Causes skin irritation.