Please login to your online account to display your discounted pricing

Cerium(III) nitrate hexahydrate, 99.99% (REO) , Alfa Aesar™, REacton™

Chemical Identifiers

Chemical Name or Material Cerium(III) nitrate
CAS 10294-41-4
Physical Form Crystalline Aggregates
Assay Percent Range 99.99% (REO)
Molecular Formula Ce(NO3)3·6H2O
Formula Weight 434.22; 326.13 (anhydrous)
MDL Number MFCD00149631
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1133018

  SDS
alfa aesar
1133018
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1133006

  SDS
alfa aesar
1133006
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1133030

  SDS
alfa aesar
1133030
250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 434.23 (326.13 Anhydrous)
Quantity 50g
Name Note hexahydrate
Storage Note 1 Hygroscopic
Physical Form Crystalline Aggregates
EINECS Number 233-297-2
Assay Percent Range 99.99% (REO)
Molecular Formula CeN3O9.6H2O N5Na2O
Chemical Name or Material Cerium(III) nitrate hexahydrate
CAS 10294-41-4
MDL Number MFCD00149631
Safety and Handling

Hygroscopic