Please login to your online account to display your discounted pricing

Cerium(III) chloride, ultra dry, 99.9% (REO), Alfa Aesar™

Chemical Identifiers

Chemical Name or Material Cerium(III) chloride
CAS 7790-86-5
Name Note ultra dry, (REO)
Molecular Formula CeCl3
Formula Weight 246.48
MDL Number MFCD00010929
Merck Index 14,1997
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AA3568814

 
alfa aesar
3568814
Ampouled under Argon 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 246.48
Quantity 25g
Chemical Name or Material Cerium(III) chloride
TSCA TSCA
Odor Odorless
Packaging Ampouled under Argon
Assay 99.9%
Sensitivity Hygroscopic
Molecular Formula CeCl3
MDL Number MFCD00010929
CAS 7790-86-5
Merck Index 14,1997
Boiling Point 1727°C
Color White to Pale Gray
Melting Point 848°C
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number FK5075000
Name Note ultra dry, (REO)
Physical Form Crystalline
Density 3.92
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.