Please login to your online account to display your discounted pricing

Cerium(III) chloride, ultra dry, 99.9% (REO), Alfa Aesar

$74.28 - $729.27

Chemical Identifiers

Chemical Name or Material Cerium(III) chloride
CAS 7790-86-5
Purity Grade Ultra dry
Physical Form Crystalline
Assay Percent Range 99.9%
Molecular Formula CeCl3
Formula Weight 246.48
MDL Number MFCD00010929
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3568806

  SDS
alfa aesar
3568806
5g Each for $74.28

AA3568822

  SDS
alfa aesar
3568822
100g Each for $729.27
Description & Specifications

Specifications

Formula Weight 246.48
Quantity 5g
Color White
Density 3.92g/cm3 at 20°C
Chemical Name or Material Cerium(III) chloride
EINECS Number 232-227-8
Purity Grade Ultra dry
Physical Form Crystalline
Health Hazard 1 H315-H319-H335 : Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
Odor Odorless
Assay Percent Range 99.9%
Molecular Formula CeCl3
MDL Number MFCD00010929
CAS 7790-86-5
Appearance White crystalline beads
Storage Note 1 Hygroscopic
Safety and Handling

H315-H319-H335 : Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

Hygroscopic