missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cerium(III) chloride heptahydrate, 99%, ACROS Organics™

Manufacturer:  Fisher Scientific Acros AC211820250

 View more versions of this product

Catalog No. AC211820250



Specifications

Specifications

Cerium(III) chloride heptahydrate
18618-55-8
CeCl3·7H2O
MFCD00149634
10, 1956
372.58
Crystals or Crystalline Powder
3.97
Solubility in water: 1000g/L (20°C). Other solubilities: soluble in alcohol
848.0°C
99%
98.5% min. (Argentometry)
CeCl3·7H2O
01,121; 09,99; 15,72; 16,4; 17,314
Solubility in water: 1000g/L (20°C). Other solubilities: soluble in alcohol
372.58
99%
Glass bottle
Yellow
25g

Chemical Identifiers

18618-55-8
CeCl3·7H2O
372.58
MFCD00149634
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
Very toxic to aquatic life with long lasting effects.

GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective

GHS Signal Word: Danger

SDS Documents