Learn More
Cerium(III) chloride heptahydrate, 99%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 211825000
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Cerium(III) chloride heptahydrate | |
18618-55-8 | |
98.5% min. (Argentometry) | |
CeCl3·7H2O | |
01,121; 09,99; 15,72; 16,4; 17,314 | |
Solubility in water: 1000g/L (20°C). Other solubilities: soluble in alcohol | |
372.58 | |
Glass bottle |
99% | |
500g | |
CeCl3·7H2O | |
MFCD00149634 | |
10, 1956 | |
372.58 | |
99% | |
3.97 |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Very toxic to aquatic life with long lasting effects.
GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective
GHS Signal Word: Danger
RUO â Research Use Only