missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cerium(III) chloride, anhydrous, 99.5% (REO), Alfa Aesar™

For Incandescent Gas Mantles, Spectrography, Polymerization of Olefins

Manufacturer:  Alfa Aesar 2111230

 View more versions of this product

Catalog No. AA2111230



Specifications

Specifications

Cerium(III) chloride
99.5%
MFCD00010929
cerium iii chloride, cerous chloride, cerium iii chloride, cerium iii chloride, anhydrous, cerium-144 chloride, cerium chloride generic, cerium-chloride, cerium chloride octahydrate, cerous iii-chloride, cerium iii chloride
[Cl-].[Cl-].[Cl-].[Ce+3]
246.466
CHEBI:35458
Powder
White
7790-86-5
CeCl3
White powder
VYLVYHXQOHJDJL-UHFFFAOYSA-K
cerium(3+);trichloride
24636
246.48
Odorless
250g

Chemical Identifiers

7790-86-5
246.466
VYLVYHXQOHJDJL-UHFFFAOYSA-K
24636
cerium(3+);trichloride
CeCl3
MFCD00010929
cerium iii chloride, cerous chloride, cerium iii chloride, cerium iii chloride, anhydrous, cerium-144 chloride, cerium chloride generic, cerium-chloride, cerium chloride octahydrate, cerous iii-chloride, cerium iii chloride
CHEBI:35458
[Cl-].[Cl-].[Cl-].[Ce+3]
Safety and Handling

Safety and Handling

H315-H319-H335 : Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

EINECSNumber : 232-227-8

SDS Documents