Learn More
Cerium(III) chloride, 99.9%, pure, anhydrous, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 369750050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
1727.0°C | |
848.0°C | |
7790-86-5 | |
Powder | |
99.90% | |
CeCl3 | |
15, 1996 | |
Solubility in water: soluble | |
[Cl-].[Cl-].[Cl-].[Ce+3] | |
246.48 | |
CHEBI:35458 | |
99.9% | |
Glass bottle |
Cerium(III) chloride | |
Anhdrous | |
White to Gray | |
5 g | |
CeCl3 | |
MFCD00010929 | |
cerium iii chloride, cerous chloride, cerium iii chloride, anhydrous, cerium-144 chloride, cerium chloride generic, cerium-chloride, cerium chloride octahydrate, cerous iii-chloride | |
VYLVYHXQOHJDJL-UHFFFAOYSA-K | |
cerium(3+);trichloride | |
24636 | |
246.48 | |
Pure |
Chemical Identifiers
7790-86-5 | |
246.48 | |
VYLVYHXQOHJDJL-UHFFFAOYSA-K | |
24636 | |
cerium(3+);trichloride |
CeCl3 | |
MFCD00010929 | |
cerium iii chloride, cerous chloride, cerium iii chloride, anhydrous, cerium-144 chloride, cerium chloride generic, cerium-chloride, cerium chloride octahydrate, cerous iii-chloride | |
CHEBI:35458 | |
[Cl-].[Cl-].[Cl-].[Ce+3] |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Very toxic to aquatic life with long lasting effects.
GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective
GHS Signal Word: Danger
EINECSNumber : 232-227-8
RUO – Research Use Only