missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Cerium(III) carbonate hydrate, REacton™, 99.999% (REO)

Manufacturer:  Alfa Aesar™ 4449636

 View more versions of this product

Catalog No. AA4449636



Specifications

Specifications

Cerium (III) carbonate hydrate
(REO)
MFCD00149635
acmc-20alhm
KHSBAWXKALEJFR-UHFFFAOYSA-H
cerium(3+);tricarbonate;hydrate
16212049
Crystalline
Reacton™
500g
54451-25-1
C3H2Ce2O10
14,1996
Soluble in dilute mineral acids. Insoluble in water.
C(=O)([O-])[O-].C(=O)([O-])[O-].C(=O)([O-])[O-].O.[Ce+3].[Ce+3]
478.271
460.27 (Anhydrous)
99.999%
848°C

Chemical Identifiers

54451-25-1
478.271
KHSBAWXKALEJFR-UHFFFAOYSA-H
16212049
C(=O)([O-])[O-].C(=O)([O-])[O-].C(=O)([O-])[O-].O.[Ce+3].[Ce+3]
C3H2Ce2O10
MFCD00149635
acmc-20alhm
cerium(3+);tricarbonate;hydrate
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 208-655-6

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents