Please login to your online account to display your discounted pricing

Cerium(III) carbonate hydrate, 99.9%, ACROS Organics™


Quantity 250g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC379402500

 View more versions of this product

Catalog No. AC379402500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Purity Grade Trace Metal Basis
Physical Form Crystalline Powder
Formula Weight 460.27
Quantity 250g
Color White to off-white
Linear Formula Ce(CO3)3·xH2O
Chemical Name or Material Cerium(III) carbonate hydrate
Packaging Glass bottle
Assay Percent Range 99.9%
Molecular Formula C3Ce2O9·xH2O
Total Trace Metal Impurities 0.1% max.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
MDL Number MFCD00149635
CAS 54451-25-1
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

Specifications

Chemical Name or Material Cerium(III) carbonate hydrate
CAS 54451-25-1
Purity Grade Trace Metal Basis
Assay Percent Range 99.9%
Molecular Formula C3Ce2O9·xH2O
Linear Formula Ce(CO3)3·xH2O
Formula Weight 460.27
MDL Number MFCD00149635
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

SDS & Safety Documents