Learn More
Cerium(III) bromide, ultra dry, 99.99% (metals basis), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 04719414
Description
It is used as an additive for chemical reagent. Undoped single crystals of CeBr3 have shown promise as a γ-ray scintillation detector in nuclear non-proliferation testing, medical imaging, environmental remediation, and oil exploration.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Ampouled under argon
Specifications
1560°C | |
733°C | |
14457-87-5 | |
25 g | |
Br3Ce | |
14,1995 | |
MOOUSOJAOQPDEH-UHFFFAOYSA-K | |
cerium(3+) tribromide | |
379.83 | |
Odorless | |
Hygroscopic |
Cerium(III) bromide | |
Ultra dry | |
Beads | |
(metals basis) | |
MFCD00016004 | |
Soluble in water. | |
[Br-].[Br-].[Br-].[Ce+3] | |
379.83 | |
99.99% | |
Ampule | |
5.18 g/mL |
Chemical Identifiers
14457-87-5 | |
379.83 | |
MOOUSOJAOQPDEH-UHFFFAOYSA-K | |
[Br-].[Br-].[Br-].[Ce+3] |
Br3Ce | |
MFCD00016004 | |
cerium(3+) tribromide |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 238-447-0
TSCA : Yes
storageNote1 :
- Ampouled under argon
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only