Learn More
Cerium(III) bromide hydrate, 99.998% (metals basis), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 04443209
Description
Cerium(III) bromide is used in the following ways, CeBr3-doped lanthanum bromide single crystals are known to exhibit superior scintillation properties for applications in the security, medical imaging, and geophysics detectors.Undoped single crystals of CeBr3 have shown promise as a γ-ray scintillation detector in nuclear non-proliferation testing, medical imaging, environmental remediation, and oil exploration
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Cerium (III) bromide hydrate | |
14457-87-5 | |
10 g | |
Br3Ce | |
14,1995 | |
MOOUSOJAOQPDEH-UHFFFAOYSA-K | |
cerium(3+) tribromide | |
379.83 (Anhydrous) | |
Odorless |
Orange | |
Crystalline | |
(metals basis) | |
MFCD00016004 | |
Soluble in water. | |
[Br-].[Br-].[Br-].[Ce+3] | |
379.83 | |
99.998% |
Chemical Identifiers
14457-87-5 | |
379.83 | |
MOOUSOJAOQPDEH-UHFFFAOYSA-K | |
[Br-].[Br-].[Br-].[Ce+3] |
Br3Ce | |
MFCD00016004 | |
cerium(3+) tribromide |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 238-447-0
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only