Learn More
Cerium(III) bromide hydrate, 99%, Thermo Scientific Chemicals
$277.78 - $976.59
Chemical Identifiers
CAS | 14457-87-5 |
---|---|
Molecular Formula | Br3Ce |
Molecular Weight (g/mol) | 379.83 |
MDL Number | MFCD00016004 |
InChI Key | MOOUSOJAOQPDEH-UHFFFAOYSA-K |
IUPAC Name | cerium(3+) tribromide |
SMILES | [Br-].[Br-].[Br-].[Ce+3] |
Description
Cerium(III) bromide hydrate is used in the security, medical imaging, and geophysics detectors, γ-ray scintillation detector in nuclear non-proliferation testing, medical imaging, environmental remediation, and oil exploration.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
14457-87-5 | |
379.83 | |
MOOUSOJAOQPDEH-UHFFFAOYSA-K | |
[Br-].[Br-].[Br-].[Ce+3] |
Br3Ce | |
MFCD00016004 | |
cerium(3+) tribromide |
Specifications
14457-87-5 | |
25 g | |
MFCD00016004 | |
MOOUSOJAOQPDEH-UHFFFAOYSA-K | |
cerium(3+) tribromide | |
379.83 (Anhydrous) | |
Odorless |
Crystalline | |
Br3Ce | |
Soluble in water. | |
[Br-].[Br-].[Br-].[Ce+3] | |
379.83 | |
99% | |
Cerium (III) bromide hydrate |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 238-447-0
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only