missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Cerium(III) bromide hydrate, 99%

Manufacturer:  Alfa Aesar™ 4444714

 View more versions of this product

Catalog No. AA4444714



Specifications

Specifications

Cerium (III) bromide hydrate
CeBr3·xH2O
Soluble in water.
Crystalline
Odorless
14457-87-5
MFCD00150206
379.83 (Anhydrous)
99%
25g

Chemical Identifiers

14457-87-5
CeBr3·xH2O
MFCD00150206
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 238-447-0

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents