missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Cerium(III) acetate sesquihydrate, REacton™, 99.99% (REO)

Manufacturer:  Alfa Aesar™ 1132418

 View more versions of this product

Catalog No. AA1132418



Specifications

Specifications

Cerium(III) acetate sesquihydrate
(REO)
MFCD00051060
Soluble in water (265g/L) at 15°C.
CC(=O)[O-].CC(=O)[O-].CC(=O)[O-].[Ce+3]
317.248
344.28 (317.26 Anhydrous)
99.99%
308°C (decomposition)
537-00-8
C6H9CeO6
acetic acid, cerium 3+ salt, cerium 3+ acetate, cerium 3+ triacetate, cerium acetate, cerium iii acetate, cerium iii acetate sesquihydrate, cerium triacetate, cerous acetate, unii-yyb5291r2h
VGBWDOLBWVJTRZ-UHFFFAOYSA-K
cerium(3+);triacetate
10826
Crystalline Aggregates
Reacton™
50g

Chemical Identifiers

537-00-8
317.248
VGBWDOLBWVJTRZ-UHFFFAOYSA-K
10826
CC(=O)[O-].CC(=O)[O-].CC(=O)[O-].[Ce+3]
C6H9CeO6
MFCD00051060
acetic acid, cerium 3+ salt, cerium 3+ acetate, cerium 3+ triacetate, cerium acetate, cerium iii acetate, cerium iii acetate sesquihydrate, cerium triacetate, cerous acetate, unii-yyb5291r2h
cerium(3+);triacetate
Safety and Handling

Safety and Handling

GHS H Statement
H318-H335-H315
Causes serious eye damage.
May cause respiratory irritation.
Causes skin irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P310-P312-P332+P313-P362-P501c

H315-H318-H335

EINECSNumber : 208-654-0

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents