missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Cerium(III) acetate sesquihydrate, 99.9% (REO)

With bromide ion, catalyzes the liquid-phase auto-oxidation of cresols

Manufacturer:  Alfa Aesar™ 1529422

 View more versions of this product

Catalog No. AA1529422



Specifications

Specifications

Cerium(III) acetate sesquihydrate
(REO)
MFCD00051060
Soluble in water
CC(=O)[O-].CC(=O)[O-].CC(=O)[O-].[Ce+3]
317.248
344.28 (317.26 Anhydrous)
99.9%
308°C (decomposition)
537-00-8
C6H9CeO6
cerium acetate, cerous acetate, cerium 3+ acetate, cerium iii acetate, cerium triacetate, cerium iii acetate, cerium 3+ triacetate, unii-yyb5291r2h, acetic acid, cerium 3+ salt, cerium iii acetate sesquihydrate
VGBWDOLBWVJTRZ-UHFFFAOYSA-K
cerium(3+);triacetate
10826
Powder
White
100g

Chemical Identifiers

537-00-8
317.248
VGBWDOLBWVJTRZ-UHFFFAOYSA-K
10826
CC(=O)[O-].CC(=O)[O-].CC(=O)[O-].[Ce+3]
C6H9CeO6
MFCD00051060
cerium acetate, cerous acetate, cerium 3+ acetate, cerium iii acetate, cerium triacetate, cerium iii acetate, cerium 3+ triacetate, unii-yyb5291r2h, acetic acid, cerium 3+ salt, cerium iii acetate sesquihydrate
cerium(3+);triacetate
Safety and Handling

Safety and Handling

GHS H Statement
H318-H335-H315
Causes serious eye damage.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

RTECSNumber : FK4900000

TSCA : TSCA

SDS Documents