missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cerium ICP Standard, 1mL = 10mg Ce (10,000ppm Ce), Ricca Chemical

Manufacturer:  Ricca Chemical Company RIPCE10KN100

 View more versions of this product

Catalog No. PCE10KN-100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Cerium ICP Standard
83.0,12.47,2.57
CeH12N3O15
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.[Ce+3]
434.218
Liquid
10,000ppm Ce
Colorless
10294-41-4,7697-37-2,7732-18-5
86.0,12.98,2.68
QQZMWMKOWKGPQY-UHFFFAOYSA-N
cerium(3+);trinitrate;hexahydrate
16211466
Poly natural
1 mL = 10 mg Ce
100mL

Chemical Identifiers

10294-41-4
434.218
16211466
CeH12N3O15
QQZMWMKOWKGPQY-UHFFFAOYSA-N
Documents