missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Cerium foil, 1.0mm (0.04 in.) thick, 99.9% (REO)

Manufacturer:  Alfa Aesar™ 10139FF

 View more versions of this product

Catalog No. AA10139FF



Description

Description

  • Packaged under argon
Specifications

Specifications

Cerium foil
Silver to Gray
1.0mm (0.04 in.) thick
Odorless
(REO)
MFCD00010924
cerio, ingot, cerium, chips, foil, 3n, zer, ceramics-aeium, acmc-1bl5r, chips, 3n reo, powder,-200 mesh, cerium
[Ce]
140.116
CHEBI:33369
99.9%
≈4.2g/25x25mm
25 x 25mm
7440-45-1
Mylar under Argon
Ce
UN1333
GWXLDORMOJMVQZ-UHFFFAOYSA-N
cerium
23974
Foil

Chemical Identifiers

7440-45-1
140.116
GWXLDORMOJMVQZ-UHFFFAOYSA-N
23974
cerium
Ce
MFCD00010924
cerio, ingot, cerium, chips, foil, 3n, zer, ceramics-aeium, acmc-1bl5r, chips, 3n reo, powder,-200 mesh, cerium
CHEBI:33369
[Ce]
Safety and Handling

Safety and Handling

GHS H Statement
H228-H260
Flammable solid.
In contact with water releases flammable gases which may ignite spontaneously.

GHS P Statement
P210-P231+P232-P280-P370+P378c-P402+P404-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Handle under inert gas.
Protect from moisture.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, sand, extinguishing powder.
Store in a dry place.
Store in a closed container.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: CERIUM

TSCA : TSCA

storageNote1 :

  • Packaged under argon

Documents