missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cerium foil, 0.1mm (0.004in) thick, 99.9% (REO), Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 010141FI

 View more versions of this product

Catalog No. AA10141FI



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
Specifications

Specifications

Silver to Gray
0.1mm (0.004 in.) thick
(REO)
MFCD00010924
50 x 50 mm
GWXLDORMOJMVQZ-UHFFFAOYSA-N
cerium
23974
99.9%
Mylar under Argon
Foil
Cerium foil
7440-45-1
Ce
UN1333
cerio, ingot, cerium, chips, foil, 3n, zer, ceramics-aeium, acmc-1bl5r, chips, 3n reo, powder,-200 mesh, cerium
[Ce]
140.12
CHEBI:33369
Odorless
≈0.42g/25x25mm

Chemical Identifiers

7440-45-1
140.12
GWXLDORMOJMVQZ-UHFFFAOYSA-N
23974
cerium
Ce
MFCD00010924
cerio, ingot, cerium, chips, foil, 3n, zer, ceramics-aeium, acmc-1bl5r, chips, 3n reo, powder,-200 mesh, cerium
CHEBI:33369
[Ce]
Safety and Handling

Safety and Handling

GHS H Statement
H228-H260
Flammable solid.
In contact with water releases flammable gases which may ignite spontaneously.

P210-P240-P241-P280-P370+P378q

H228

DOTInformation : Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: CERIUM

EINECSNumber : 231-154-9

TSCA : Yes

storageNote1 :

  • Packaged under argon

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only