missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cerium, AAS standard solution, Specpure™ Ce 1000μg/mL, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 030520AP

 View more versions of this product

Catalog No. AA30520AP



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Cerium AAS standard solution
Ce 1000μg/mL
MFCD00010924
Specpure™
Ce in 5% HNO3
500 mL
UN3264
Liquid

Chemical Identifiers

MFCD00010924
Safety and Handling

Safety and Handling

GHS H Statement
H314
Causes severe skin burns and eye damage.

P234-P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362-P390

H290-H315-H319

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, INORGANIC, N.O.S.

EINECSNumber : 231-714-2

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only