Please login to your online account to display your discounted pricing

Cerium AA Standard, 1mL = 1mg Ce (1,000ppm Ce), Ricca Chemical


Quantity 500mL
Packaging Poly natural

Manufacturer: ricca chemical company  RIACE1KN500

 View more versions of this product

Catalog No. ACE1KN-500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Liquid
Color Clear colorless
Quantity 500mL
Chemical Name or Material Cerium AA Standard
CAS 7732-18-5
Concentration or Composition Notes 1,000ppm Ce
CAS Max % 0.3
CAS Min % 93.0
CAS Max % 97.0
CAS 7697-37-2
Packaging Poly natural
CAS Min % 4.21
CAS Max % 4.39
CAS 10294-41-4
Equivalence 1 mL = 1 mg Ce
CAS Min % 0.29

Specifications

Chemical Name or Material Cerium AA Standard
CAS 7732-18-5
CAS Min % 93.0
CAS Max % 97.0
CAS 7697-37-2
CAS Min % 4.21
CAS Max % 4.39
CAS 10294-41-4
CAS Min % 0.29
CAS Max % 0.3
Physical Form Liquid