Please login to your online account to display your discounted pricing

Ceric Ammonium Nitrate ACS AR, Macron Fine Chemicals™


macron

Quantity 125g
Packaging POLYSTORMOR

Manufacturer: avantor performance  256801

 View more versions of this product

Catalog No. MK2568125

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 548.23
Purity Grade ACS Grade
Chemical Name or Material Ceric Ammonium Nitrate
CAS 16774-21-3
Packaging POLYSTORMOR
Molecular Formula (NH4)2Ce(NO3)6
Quantity 125g

Specifications

Chemical Name or Material Ceric Ammonium Nitrate
CAS 16774-21-3
Purity Grade ACS Grade
Molecular Formula (NH4)2Ce(NO3)6
Formula Weight 548.23