missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ceric Ammonium Nitrate ACS AR, Macron Fine Chemicals™

macron

Chemical Identifiers

CAS 16774-21-3
Molecular Formula (NH4)2Ce(NO3)6
Formula Weight 548.23
Chemical Name or Material Ceric Ammonium Nitrate
Grade ACS Grade
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
MK2568125 SDS Avantor Performance
256801
POLYSTORMOR 125g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
MK2568500 Avantor Performance
256804
POLYSTORMOR 500g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

16774-21-3
548.23
ACS Grade
(NH4)2Ce(NO3)6
Ceric Ammonium Nitrate

Specifications

548.23
POLYSTORMOR
125g
ACS Grade
16774-21-3
(NH4)2Ce(NO3)6
Ceric Ammonium Nitrate